Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Allis Tissue Forceps, Sklar, 9-1/2 in., 5x6 Teeth, Straight, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902279
Sklar Black Allis Tissue Forceps, Sklar, 9-1/2 in., 5x6 Teeth, Straight, Stainless Steel with Reduced Reflectivity Polymer Coating. Sklar Black Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue during laser surgery. They are commonly used with tonsil, vaginal, breast, and thyroid tissue. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.
Catalog No. 50-143-6565