Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
315-30-0
C5H4N4O
136.114
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
allopurinol,1h-pyrazolo 3,4-d pyrimidin-4-ol,zyloprim,zyloric,lopurin,atisuril,bleminol,caplenal,uripurinol,embarin
2094
CHEBI:40279
1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
C1=C2C(=NC=NC2=O)NN1
315-30-0
C5H4N4O
136.114
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
allopurinol,1h-pyrazolo 3,4-d pyrimidin-4-ol,zyloprim,zyloric,lopurin,atisuril,bleminol,caplenal,uripurinol,embarin
2094
CHEBI:40279
1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
C1=C2C(=NC=NC2=O)NN1
Quantity 250g
Color White-Yellow
Melting Point 384°C
UN Number 2811
Formula Weight 136.11
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Allopurinol

RTECSNumber : UR0785000

TSCA : Yes