Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog Number 18600405
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
315-30-0
C5H4N4O
136.11
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12
315-30-0
C5H4N4O
136.11
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12

A1577, 315-30-0, C5H4N4O

Spectrum™ Chemical Allopurinol, USP is a xanthine oxidase inhibitor that is primarily used to treat Hyperuricemia. It is on the World Health Organization′s List of Essential Medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
UN Number UN2811
Percent Purity 98 to 102%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit