Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allopurinol, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
315-30-0
C5H4N4O
136.11
OFCNXPDARWKPPY-UHFFFAOYSA-N
1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12
315-30-0
C5H4N4O
136.11
OFCNXPDARWKPPY-UHFFFAOYSA-N
1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12

A1577, 315-30-0, C5H4N4O

Spectrum™ Allopurinol, USP is a xanthine oxidase inhibitor that is primarily used to treat Hyperuricemia. It is on the World Health Organization′s List of Essential Medicines. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 100 g
Loss on Drying 0.005
CAS Max % 1
UN Number UN2811
Percent Purity 98 to 102%