Learn More
Humboldt Alloy Tool Steel Mortar and Pestle Set
Plattner's diamond mortar and pestle set for crushing small ore samples
Manufacturer:  Humboldt H17270
Catalog No. 12-990
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Mortar has flat-bottomed cylindrical cavity
  • Fitted with removable sleeve for retaining crushed material in cavity
  • Pestle fits snugly inside sleeve
Order Info
Cavity, 1 dia. × 1/4 in. deep (25 × 6.4mm). Mortar O.D., 3 in. (76mm). Pestle diameter, 1 in. (25mm). Pestle length, 3 in. (76mm).
Specifications
Specifications
3 in.
Mortar and Pestle
For crushing small samples of ore
7.6cm
Steel