Learn More
Grace™ Alltech™ Combo Packed GC Columns
Specifications
Material Stainless Steel
Length (Metric) 1.8 m
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
AT12430PC
View Documents
Grace™
5124848
N/A SS GC Packed Column; 6 ft. x 0.125 in. O.D.; 10% Silar 10C on Chromosorb WHP 100/120 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Part Not Found
N/A
Description
Description
Specifications
Specifications
Stainless Steel
1.8 m