Learn More
Grace™ Alltech™ Denali™ C18 Reversed Phase Columns
Manufacturer:  Grace™ 5112970
 View more versions of this product
Catalog No. A238DE10415
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
0.5 to 1.5mL/min.
Spherical
C18
L1
20%
120Å
5μm
238DE C18
Denali™ C18 reversed phase column
280 to 340m2/g
HPLC
150mm
Yes
4.6mm