Learn More
Grace™ Alltech™ Vydac™ 208TP C8 Column
Excellent peak shape with little or no TFA
Manufacturer:  Grace™ 5103849
 View more versions of this product
Catalog No. 208TP152022
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
208TP C8
Vydac™ TP C8 column
60 to 110m2/g
HPLC
250mm
Yes
22mm
Spherical
208TP C8
L7
5%
300Å
15 to 20μm