Grace™ Alltech™ Vydac™ 218TP C18 Columns and Guard Cartridges

Well suited for microbore applications, stable in popular microbore mobile phases

Manufacturer: Grace™ 5103970

 View more versions of this product

Catalog No. 218TP101510

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Particle Shape Spherical
    Type HPLC Column
    Stationary Phase 218TP
    Surface Area 60 to 110m2/g
    Temperature 50°C
    USP Type L1
    Column Type C18
    Carbon Load 8%
    Length (Metric) 250mm
    Pore Size 300Å
    Endcapped Yes
    Particle Size 10 to 15μm
    Diameter (Metric) Inner 10mm
    Flow Rate 1.2mL/min.
    For Use With Separation of small polypeptides <4000 to 5000 M.W., enzymatic digest fragments, natural and synthetic Peptides, multi-ring compounds
    Packing Material Silica
    Mobile Phase Compatibility A : 25% ACN/0.1% TFA in Water, B : 58% ACN/0.1% TFA in Water

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit