Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grace™ Alltech™ Vydac™ 218TP C18 Columns and Guard Cartridges

Well suited for microbore applications, stable in popular microbore mobile phases

Manufacturer:  Grace™ 5103976

 View more versions of this product

Catalog No. A218TP10415

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

1.2mL/min.
Silica
Spherical
218TP
50°C
C18
150mm
Yes
4.6mm
Separation of small polypeptides <4000 to 5000 M.W., enzymatic digest fragments, natural and synthetic Peptides, multi-ring compounds
A : 25% ACN/0.1% TFA in Water, B : 58% ACN/0.1% TFA in Water
HPLC Column
60 to 110m2/g
L1
8%
300Å
10μm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit