Learn More
Grace™ Alltech™ Vydac™ 218TP C18 Columns and Guard Cartridges
Well suited for microbore applications, stable in popular microbore mobile phases
Manufacturer:  Grace™ 5104329
 View more versions of this product
Catalog No. A218TP10154
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
1.2mL/min.
C18
250mm
10-15μm
Separation of small polypeptides <4000 to 5000 M.W., enzymatic digest fragments, natural and synthetic Peptides, multi-ring compounds
8%
Yes
4.6mm