Grace™ Alltech™ Vydac™ 218TP C18 Columns and Guard Cartridges

Well suited for microbore applications, stable in popular microbore mobile phases

Manufacturer: Grace™ 5103977

 View more versions of this product

Catalog No. 218TP152022

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Flow Rate 1.2mL/min.
    For Use With Separation of small polypeptides <4000 to 5000 M.W., enzymatic digest fragments, natural and synthetic Peptides, multi-ring compounds
    Column Type C18
    Carbon Load 8%
    Length (Metric) 250mm
    Endcapped Yes
    Particle Size 15-20μm
    Diameter (Metric) Inner 22mm

    • Acid and base deactivated symmetrical peaks
    • Polymerically bonded and double-endcapped for long column lifetimes
    • Analyze acids, bases, and neutrals in one run

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit