Alluviam HazMasterG3™ CBRNE/IED Threat Identification System and HazMapper™ Threat-Mapping Software
Choose between comprehensive mobile HazMat/CBRNE/IED decision aid and threat-mapping software, or integrate the two
Manufacturer: Alluviam LLCSupplier Diversity Partner HMG3DPPC
 View more versions of this product
Catalog No. 19-130-6002
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product.
Please contact customer service for assistance: 1-800-766-7000.
Add to Cart
    Specifications
    Certifications/Compliance U.S.DHS, DoD COE/JTA
    Includes Software for handheld computing devices comes on SD/MMC memory card with CD-ROM backup, CD-ROM contains program files, user manual and includes 37+ responder reference guides, first year of annual maintenance and support
    For Use With (Application) Identify an extremely broad range of unknown chemical agents, radiological isotopes, complex improvised explosive device (IED) threats, detonators, intermodal containers and road and rail hazards
    Product Type Handheld pocket PC device
  • Description & Specifications
Related Products
Product Content Feedback