Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl acetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
591-87-7
C5H8O2
100.117
MFCD00008721
FWZUNOYOVVKUNF-UHFFFAOYSA-N
allyl acetate,3-acetoxypropene,acetic acid, 2-propenyl ester,2-propenyl acetate,2-propenyl ethanoate,3-acetoxy-1-propene,allylacetate,acetic acid, allyl ester,2-propenyl methanoate,unii-e4u5e5990i
11584
prop-2-enyl acetate
CC(=O)OCC=C
591-87-7
C5H8O2
100.117
MFCD00008721
FWZUNOYOVVKUNF-UHFFFAOYSA-N
allyl acetate,3-acetoxypropene,acetic acid, 2-propenyl ester,2-propenyl acetate,2-propenyl ethanoate,3-acetoxy-1-propene,allylacetate,acetic acid, allyl ester,2-propenyl methanoate,unii-e4u5e5990i
11584
prop-2-enyl acetate
CC(=O)OCC=C
Quantity 5mL
Infrared Spectrum Authentic
Color Colorless
Refractive Index 1.4030 to 1.4050 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CO2CH2CH=CH2
Beilstein 02, 136
Merck Index 15, 276
Formula Weight 100.12
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Allyl acetate, 99%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-734-8

RTECSNumber : AF1750000

TSCA : TSCA