Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl acetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
591-87-7
C5H8O2
100.12
MFCD00008721
FWZUNOYOVVKUNF-UHFFFAOYSA-N
allyl acetate,3-acetoxypropene,acetic acid, 2-propenyl ester,2-propenyl acetate,2-propenyl ethanoate,3-acetoxy-1-propene,allylacetate,acetic acid, allyl ester,2-propenyl methanoate,unii-e4u5e5990i
11584
prop-2-enyl acetate
CC(=O)OCC=C
591-87-7
C5H8O2
100.12
MFCD00008721
FWZUNOYOVVKUNF-UHFFFAOYSA-N
allyl acetate,3-acetoxypropene,acetic acid, 2-propenyl ester,2-propenyl acetate,2-propenyl ethanoate,3-acetoxy-1-propene,allylacetate,acetic acid, allyl ester,2-propenyl methanoate,unii-e4u5e5990i
11584
prop-2-enyl acetate
CC(=O)OCC=C
Color Colorless
Specific Gravity 0.92
Infrared Spectrum Authentic
Density 0.9200g/mL
Refractive Index 1.4030 to 1.4050
Packaging Glass bottle
Quantity 100mL
Boiling Point 103.0°C to 104.0°C
Melting Point -96.0°C
Flash Point 6°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CO2CH2CH=CH2
Beilstein 02, 136
Merck Index 15, 276
Solubility Information Solubility in water: insoluble
Formula Weight 100.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Allyl acetate, 99%
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin

GHS Signal Word: Danger

EINECSNumber : 209-734-8

RTECSNumber : AF1750000

TSCA : TSCA