Alfa Aesar™ Allyl acrylate, 95%, stab. with 4-methoxyphenol

Manufacturer: Alfa Aesar™ 4446322

Quantity 100g

 View more versions of this product

Catalog No. AA4446322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 999-55-3
    Molecular Formula CH2=CHCOOCH2CH=CH2
    Formula Weight 112.13
    MDL Number MFCD00014949
    UN Number UN2929
    Beilstein 1743576
    Chemical Name or Material Allyl acrylate, Stabilized with 4-methoxyphenol

    Specifications

    Formula Weight 112.13
    Health Hazard 2 GHS H Statement
    H310-H330-H226-H302-H315-H319-H335
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Refractive Index 1.4360
    Boiling Point 46° to 47°C (40mmHg)
    RTECS Number UD3675000
    TSCA TSCA
    Molecular Formula CH2=CHCOOCH2CH=CH2
    MDL Number MFCD00014949
    Density 0.935g/mL
    Beilstein 1743576
    Quantity 100g
    CAS 999-55-3
    UN Number UN2929
    Health Hazard 1 Danger
    Flash Point 32°C (89°F)
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
    Odor Pungent
    Chemical Name or Material Allyl acrylate, Stabilized with 4-methoxyphenol
    Assay 95%

    GHS H Statement
    H310-H330-H226-H302-H315-H319-H335
    Fatal in contact with skin.
    Fatal if inhaled.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit