Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl chloroacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass Bottle
2916-14-5
C5H7ClO2
134.56
MFCD00058936
VMBJJCDVORDOCF-UHFFFAOYSA-N
allyl chloroacetate,allyl 2-chloroacetate,chloroacetic acid allyl ester,acetic acid, chloro-, 2-propenyl ester,prop-2-en-1-yl 2-chloroacetate,acetic acid, chloro-, allyl ester,allyl monochloroacetate,acmc-1cffa,chloroacetic acid,2-propenyl ester
76206
prop-2-enyl 2-chloroacetate
C=CCOC(=O)CCl
2916-14-5
C5H7ClO2
134.56
MFCD00058936
VMBJJCDVORDOCF-UHFFFAOYSA-N
allyl chloroacetate,allyl 2-chloroacetate,chloroacetic acid allyl ester,acetic acid, chloro-, 2-propenyl ester,prop-2-en-1-yl 2-chloroacetate,acetic acid, chloro-, allyl ester,allyl monochloroacetate,acmc-1cffa,chloroacetic acid,2-propenyl ester
76206
prop-2-enyl 2-chloroacetate
C=CCOC(=O)CCl
Refractive Index 1.4450 to 1.447
Boiling Point 164°C (265.0mmHg)
Density 1.1590g/mL
Flash Point 61°C
Specific Gravity 1.159
Color Colorless to Yellow
Quantity 5mL
Packaging Glass Bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 134.56
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Allyl chloroacetate, 98%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-839-8