Learn More
Allyl chloroacetate, 98%, ACROS Organics™
Specifications
Boiling Point | 164°C (265.0mmHg) |
Density | 1.1590g/mL |
Flash Point | 61°C |
Specific Gravity | 1.159 |
Refractive Index | 1.4450 to 1.447 |
Quantity | 25mL |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
Color | Colorless to Yellow |
CAS Min % | 97.5 |
Show More |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 220-839-8