Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl cumyl peroxide, Acros Organics

Click to view available options
Quantity:
1g
61808-93-3,617-94-7
C12H16O2
192.258
BKLTYOBGMZOJJS-UHFFFAOYSA-N
allyl cumyl peroxide,peroxide, 1-methyl-1-phenylethyl 2-propenyl,1-methyl-1-phenylethyl allyl peroxide,allyl-1-methyl-1-phenyl-ethyl-peroxide,2-prop-2-en-1-ylperoxy propan-2-yl benzene
2734922
2-prop-2-enylperoxypropan-2-ylbenzene
CC(C)(C1=CC=CC=C1)OOCC=C
61808-93-3,617-94-7
C12H16O2
192.258
BKLTYOBGMZOJJS-UHFFFAOYSA-N
allyl cumyl peroxide,peroxide, 1-methyl-1-phenylethyl 2-propenyl,1-methyl-1-phenylethyl allyl peroxide,allyl-1-methyl-1-phenyl-ethyl-peroxide,2-prop-2-en-1-ylperoxy propan-2-yl benzene
2734922
2-prop-2-enylperoxypropan-2-ylbenzene
CC(C)(C1=CC=CC=C1)OOCC=C
Radical initiators
Quantity 1g
Color Yellow
Density 0.95g/mL
CAS Min % 70.0,20.0
CAS Max % 80.0,30.0
Formula Weight 192.26
Physical Form Solution
Percent Purity ≥70.0%
Chemical Name or Material Allyl cumyl peroxide

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Heating may cause a fire.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger