Alfa Aesar™ Allyl ethyl ether, 98%

Manufacturer: Alfa Aesar™ B2227609

Quantity 10g

 View more versions of this product

Catalog No. AAB2227609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 557-31-3
    Molecular Formula C5H10O
    Linear Formula CH2=CHCH2OCH2CH3
    Formula Weight 86.13
    MDL Number MFCD00009255
    UN Number UN2335
    Beilstein 1361480
    Merck Index 14,293
    Synonym 3-Ethoxy-1-propene
    Chemical Name or Material Allyl ethyl ether

    Specifications

    Formula Weight 86.13
    Flash Point −20°C (−4°F)
    RTECS Number KM9120000
    TSCA TSCA
    Molecular Formula C5H10O
    Linear Formula CH2=CHCH2OCH2CH3
    MDL Number MFCD00009255
    UN Number UN2335
    Beilstein 1361480
    Merck Index 14,293
    Synonym 3-Ethoxy-1-propene
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H225-H302-H332
    Highly flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 557-31-3
    Health Hazard 1 Danger
    Boiling Point 64° to 65°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL ETHYL ETHER
    Chemical Name or Material Allyl ethyl ether
    Assay 98%

    Description

    Applications
    Allyl ethyl ether is used as an intermediate to manufacture other chemicals.

    Solubility
    Soluble in organic solvents, miscible with ethanol, diethyl ether. Insoluble in water.

    Notes
    Air sensitive. Store under inert gas. Store away from oxidizing agents, air, acids.

    GHS H Statement
    H225-H302-H332
    Highly flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit