Alfa Aesar™ Allyl iodide, 97+%, stab. with copper

Manufacturer: Alfa Aesar™ A1182230

Quantity 250g

 View more versions of this product

Catalog No. AAA1182230

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 556-56-9
    Molecular Formula C3H5I
    Linear Formula CH2=CHCH2I
    Formula Weight 167.98
    MDL Number MFCD00001094
    UN Number UN1723
    Beilstein 1697594
    Merck Index 14,294
    Synonym 3-Iodo-1-propene
    Chemical Name or Material Allyl iodide, Stabilized with copper

    Specifications

    Formula Weight 167.98
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula CH2=CHCH2I
    Boiling Point 101° to 103°C
    Molecular Formula C3H5I
    Flash Point 18°C (64°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALLYL IODIDE
    Melting Point −99°C
    CAS 556-56-9
    Beilstein 1697594
    Recommended Storage 2°C to 8°C
    TSCA TSCA
    MDL Number MFCD00001094
    Quantity 250g
    Synonym 3-Iodo-1-propene
    UN Number UN1723
    Merck Index 14,294
    RTECS Number UD0450000
    Chemical Name or Material Allyl iodide, Stabilized with copper
    Assay ≥97%

    Description

    Applications
    Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

    Solubility
    Soluble in alcohol, chloroform, ether, and diethyl ether. Insoluble in water.

    Notes
    Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with heat, light, amines, nitrides, azo/diazo compounds, alkali metals, and epoxides.

    GHS H Statement
    H225-H314
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.

    2°C to 8°C

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit