Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals

Catalog Number p-52127
Click to view available options
Quantity:
25 g
100 g
870-23-5
C3H6S
74.14
MFCD00004894
ULIKDJVNUXNQHS-UHFFFAOYSA-N
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol
13367
prop-2-ene-1-thiol
SCC=C
870-23-5
C3H6S
74.14
MFCD00004894
ULIKDJVNUXNQHS-UHFFFAOYSA-N
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol
13367
prop-2-ene-1-thiol
SCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 67°C to 68°C
Flash Point 18°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4745 to 1.4785
Linear Formula H2C=CHCH2SH
Quantity 25 g
Beilstein 01, 440
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 74.13
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material Allyl mercaptan
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 212-792-7
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit