Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl mercaptan, tech., 70%, remainder mainly sulfide, Thermo Scientific Chemicals

Click to view available options
Quantity:
25 g
100 g
870-23-5
C3H6S
74.13
MFCD00004894
ULIKDJVNUXNQHS-UHFFFAOYSA-N
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol
13367
prop-2-ene-1-thiol
C=CCS
870-23-5
C3H6S
74.13
MFCD00004894
ULIKDJVNUXNQHS-UHFFFAOYSA-N
allyl mercaptan, 2-propene-1-thiol, allylthiol, allylmercaptan, allyl sulfhydrate, unii-1x587iby09, 2-propenyl mercaptan, 2-propenyl-1-thiol, ch2=chch2sh, allyl thiol
13367
prop-2-ene-1-thiol
C=CCS

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 67°C to 68°C
Flash Point 18°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4745 to 1.4785
Linear Formula H2C=CHCH2SH
Quantity 100 g
Beilstein 01, 440
Specific Gravity 0.93
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 74.13
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material Allyl mercaptan
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Danger

EINECSNumber : 212-792-7

TSCA : TSCA

RUO – Research Use Only