Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Allyl methyl sulfide, 98%

Click to view available options
Quantity:
5g
25g
10152-76-8
C4H8S
88.168
MFCD00008657
NVLPQIPTCCLBEU-UHFFFAOYSA-N
allyl methyl sulfide,1-propene, 3-methylthio,allyl methyl sulphide,3-methylthio-1-propene,sulfide, allyl methyl,methyl propenyl sulfide,unii-v7qi1r316c,methyl allyl sulfide,ccris 7066
66282
3-methylsulfanylprop-1-ene
CSCC=C
10152-76-8
C4H8S
88.168
MFCD00008657
NVLPQIPTCCLBEU-UHFFFAOYSA-N
allyl methyl sulfide,1-propene, 3-methylthio,allyl methyl sulphide,3-methylthio-1-propene,sulfide, allyl methyl,methyl propenyl sulfide,unii-v7qi1r316c,methyl allyl sulfide,ccris 7066
66282
3-methylsulfanylprop-1-ene
CSCC=C
Boiling Point 91°C to 93°C
Flash Point 18°C (65°F)
Odor Stench
Quantity 25g
UN Number UN1993
Formula Weight 88.17
Percent Purity 98%
Chemical Name or Material Allyl methyl sulfide

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C