Alfa Aesar™ Allyl n-butyl ether, 99%

Manufacturer: Alfa Aesar™ H5340814

Quantity 25g

 View more versions of this product

Catalog No. AAH5340814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3739-64-8
    Molecular Formula C7H14O
    Linear Formula CH2=CHCH2OCH2CH2CH2CH3
    Formula Weight 114.19
    MDL Number MFCD00009460
    UN Number UN3271
    Beilstein 1699588
    Chemical Name or Material Allyl n-butyl ether

    Specifications

    Formula Weight 114.19
    Molecular Formula C7H14O
    Linear Formula CH2=CHCH2OCH2CH2CH2CH3
    MDL Number MFCD00009460
    CAS 3739-64-8
    UN Number UN3271
    Quantity 25g
    Flash Point 11°C (52°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H224-H315-H319-H335
    Extremely flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
    Beilstein 1699588
    Chemical Name or Material Allyl n-butyl ether
    Assay 99%

    GHS H Statement
    H224-H315-H319-H335
    Extremely flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit