Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Phenyl Sulfone 95.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
16212-05-8
C9H10O2S
182.237
MFCD00014739
KYPIULIVYSQNNT-UHFFFAOYSA-N
allyl phenyl sulfone,benzene, 2-propenylsulfonyl,allylsulfonyl benzene,phenyl allyl sulfone,sulfone, allyl phenyl,allylsulphonyl benzene,allylphenyl sulfone,3-phenylsulfonyl-1-propene,benzene, 2-propen-1-ylsulfonyl
85330
prop-2-enylsulfonylbenzene
C=CCS(=O)(=O)C1=CC=CC=C1
16212-05-8
C9H10O2S
182.237
MFCD00014739
KYPIULIVYSQNNT-UHFFFAOYSA-N
allyl phenyl sulfone,benzene, 2-propenylsulfonyl,allylsulfonyl benzene,phenyl allyl sulfone,sulfone, allyl phenyl,allylsulphonyl benzene,allylphenyl sulfone,3-phenylsulfonyl-1-propene,benzene, 2-propen-1-ylsulfonyl
85330
prop-2-enylsulfonylbenzene
C=CCS(=O)(=O)C1=CC=CC=C1
Quantity 25g
Color Yellow
Boiling Point 128°C
Formula Weight 182.24
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material Allyl Phenyl Sulfone

RTECSNumber : WR2400000

TSCA : Yes