Alfa Aesar™ Allyl trifluoroacetate, 97%

Manufacturer: Alfa Aesar™ A1169030

Quantity 250g

 View more versions of this product

Catalog No. AAA1169030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 383-67-5
    Molecular Formula CH2=CHCH2O2CCF3
    Formula Weight 154.09
    MDL Number MFCD00013567
    UN Number UN2924
    Beilstein 1766312
    Synonym Trifluoroacetic acid allyl ester
    Chemical Name or Material Allyl trifluoroacetate

    Specifications

    Formula Weight 154.09
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula CH2=CHCH2O2CCF3
    CAS 383-67-5
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    MDL Number MFCD00013567
    Beilstein 1766312
    Synonym Trifluoroacetic acid allyl ester
    Boiling Point 73° to 75°C
    Quantity 250g
    Flash Point −1°C (30°F)
    Chemical Name or Material Allyl trifluoroacetate
    Assay 97%

    Description

    Applications
    Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.

    Solubility
    Not miscible or difficult to mix with water.

    Notes
    Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit