Alfa Aesar™ Allylchlorodimethylsilane, 97%

Manufacturer: Alfa Aesar™ B2223706

Quantity 5g

 View more versions of this product

Catalog No. AAB2223706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4028-23-3
    Molecular Formula CH2=CHCH2Si(CH3)2Cl
    Formula Weight 134.68
    MDL Number MFCD00000506
    UN Number UN2985
    Beilstein 1737978
    Synonym ADMSCl; Allyldimethylchlorosilane
    Chemical Name or Material Allylchlorodimethylsilane

    Specifications

    Formula Weight 134.68
    MDL Number MFCD00000506
    UN Number UN2985
    Quantity 5g
    Flash Point 5°C (41°F)
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
    TSCA TSCA
    Molecular Formula CH2=CHCH2Si(CH3)2Cl
    CAS 4028-23-3
    Beilstein 1737978
    Synonym ADMSCl; Allyldimethylchlorosilane
    Boiling Point 110° to 112°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Allylchlorodimethylsilane
    Assay 97%

    Description

    Applications
    Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1 ft.,4,4 ft.-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

    Solubility
    Soluble in most organic solvents; reacts with alcohols, amines, and water.

    Notes
    Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit