Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Allylchlorodimethylsilane, 97%

Manufacturer:  Alfa Aesar™ B2223714

Quantity 25g

 View more versions of this product

Catalog No. AAB2223714


  Check Availability
Add to cart

Specifications

Description

Applications
Allylchlorodimethylsilane is used in the synthesis of cyclo-1,1 ft.,4,4 ft.-bis(1,1,3,3-tetramethyl-1,3-disiloxanediyl)dibenzene, silylating agent for steroids for analysis by GC-MS. Also used in the preparation of phenylenebis(silanediyl triflates), useful synthons for organosilicon polymers, O-silylation of phenyl sulfones.

Solubility
Soluble in most organic solvents; reacts with alcohols, amines, and water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, strong bases, water/ moisture.
Description & Specifications

Chemical Identifiers

4028-23-3
134.68
UN2985
ADMSCl; Allyldimethylchlorosilane
CH2=CHCH2Si(CH3)2Cl
MFCD00000506
1737978
Allylchlorodimethylsilane

Specifications

4028-23-3
134.68
UN2985
ADMSCl; Allyldimethylchlorosilane
25g
Allylchlorodimethylsilane
CH2=CHCH2Si(CH3)2Cl
MFCD00000506
1737978
110° to 112°C
5°C (41°F)
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit