Alfa Aesar™ Allyldiphenylphosphine, 96%

Manufacturer: Alfa Aesar™ 3030309

Quantity 10g

 View more versions of this product

Catalog No. AA3030309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2741-38-0
    Linear Formula (CH2=CHCH2)P(C6H5)2
    Formula Weight 226.25
    MDL Number MFCD00014954
    UN Number UN3278
    Chemical Name or Material Allyldiphenylphosphine

    Specifications

    Formula Weight 226.25
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Flash Point >110° (230°F)
    Refractive Index 1.6190
    MDL Number MFCD00014954
    CAS 2741-38-0
    UN Number UN3278
    Health Hazard 1 Danger
    Boiling Point 194° to 200°C (15mmHg)
    Sensitivity Air Sensitive
    Linear Formula (CH2=CHCH2)P(C6H5)2
    Density 1.049g/mL
    Color Colorless
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Allyldiphenylphosphine
    Assay 96%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit