Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyldiphenylphosphine Oxide 98.0+%, TCI America™

Click to view available options
Quantity:
1g
4141-48-4
C15H15OP
242.258
MFCD00013908
PGPAPANRSWMTQO-UHFFFAOYSA-N
138131
[phenyl(prop-2-enyl)phosphoryl]benzene
C=CCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
4141-48-4
C15H15OP
242.258
MFCD00013908
PGPAPANRSWMTQO-UHFFFAOYSA-N
138131
[phenyl(prop-2-enyl)phosphoryl]benzene
C=CCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
Color White
Melting Point 113°C
Quantity 1g
Formula Weight 242.26
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Allyldiphenylphosphine Oxide