Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, ACROS Organics™
Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
Specifications
Flash Point | −40°C |
Quantity | 800mL |
Color | Gray or Green-Yellow |
Density | 0.8510g/mL |
Packaging | AcroSeal™ Glass bottle |
Specific Gravity | 0.851 |
CAS Min % | 15.37,81.22 |
CAS Max % | 18.78,84.64 |
Linear Formula | H2C=CHCH2MgBr |
UN Number | 3399 |
Show More |
Safety and Handling
GHS H Statement
Extremely flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Harmful if swallowed.
Causes severe skin bur
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 217-046-4