Learn More
Allylmagnesium chloride, 1.7M solution in THF, AcroSeal™, ACROS Organics™
Specifications
CAS Min % | 15.76,80.04 |
CAS Max % | 19.96,84.25 |
Linear Formula | H2C=CHCH2MgCl |
Concentration | 1.5 to 1.9M |
Density | 0.9600g/mL |
Formula Weight | 100.83 |
Physical Form | Liquid |
Percent Purity | 1.5 to 1.9 M |
Packaging | AcroSeal™ Glass bottle |
Solubility | Solubility in water: vigorous reaction. |
Show More |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides. In contact with water releases flammable gases which may ignite spontaneously. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger