Alfa Aesar™ Allyloxy-tert-butyldimethylsilane, 98%

Manufacturer: Alfa Aesar™ H55077AC

Quantity 25mL

 View more versions of this product

Catalog No. AAH55077AC

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 105875-75-0
    Molecular Formula C9H20OSi
    Linear Formula H2C=CHCH2OSi(CH3)2C(CH3)3
    Formula Weight 172.34
    MDL Number MFCD00239355
    UN Number UN1993
    Chemical Name or Material Allyloxy-tert-butyldimethylsilane

    Specifications

    Formula Weight 172.34
    Quantity 25mL
    Flash Point 33°C (91°F)
    Molecular Formula C9H20OSi
    Linear Formula H2C=CHCH2OSi(CH3)2C(CH3)3
    MDL Number MFCD00239355
    UN Number UN1993
    Boiling Point 120°C (50mmHg)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 105875-75-0
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Allyloxy-tert-butyldimethylsilane
    Assay 98%

    Description

    Applications
    Allyloxy-tert-butyldimethylsilane is used as a pharmaceutical intermediate.

    Solubility
    Sparingly soluble in water (0.50 g/L at 25°C).

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit