Alfa Aesar™ Allyloxytrimethylsilane, 99%

Manufacturer: Alfa Aesar™ H5344306

Quantity 5g

 View more versions of this product

Catalog No. AAH5344306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18146-00-4
    Molecular Formula C6H14OSi
    Linear Formula CH2=CHCH2OSi(CH3)3
    Formula Weight 130.26
    MDL Number MFCD00008643
    UN Number UN1993
    Beilstein 1739517
    Synonym 3-(Trimethylsiloxy)propene
    Chemical Name or Material Allyloxytrimethylsilane

    Specifications

    Formula Weight 130.26
    TSCA TSCA
    Linear Formula CH2=CHCH2OSi(CH3)3
    MDL Number MFCD00008643
    UN Number UN1993
    Quantity 5g
    Flash Point 1°C (34°F)
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Molecular Formula C6H14OSi
    CAS 18146-00-4
    Beilstein 1739517
    Synonym 3-(Trimethylsiloxy)propene
    Boiling Point 101° to 102°C
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Allyloxytrimethylsilane
    Assay 99%

    Description

    Applications
    Allyloxytrimethylsilane is used in the synthesis of metalated dienes via an isomerization-elimination sequence. Allyloxytrimethylsilane [3-(Trimethylsiloxy)propene] was used in the synthesis of trimethylsilyl protected (3-hydroxypropyl)-trimethoxysilane. Used in preparation of electrical materials, OLED, and in Pharma industry.

    Notes
    Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit