Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Allylpentafluorobenzene, 98%

Click to view available options
Quantity:
5g
25g
1736-60-3
C9H5F5
208.131
MFCD00000301
YBDBTBVNQQBHGJ-UHFFFAOYSA-N
allylpentafluorobenzene,benzene, allylpentafluoro,3-pentafluorophenyl-1-propene,2,3,4,5,6-pentafluoro allylbenzene,benzene, pentafluoro-2-propenyl,1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene,allyl pentafluorobenzene,pubchem13366,acmc-1bth5
74435
1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
C=CCC1=C(C(=C(C(=C1F)F)F)F)F
1736-60-3
C9H5F5
208.131
MFCD00000301
YBDBTBVNQQBHGJ-UHFFFAOYSA-N
allylpentafluorobenzene,benzene, allylpentafluoro,3-pentafluorophenyl-1-propene,2,3,4,5,6-pentafluoro allylbenzene,benzene, pentafluoro-2-propenyl,1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene,allyl pentafluorobenzene,pubchem13366,acmc-1bth5
74435
1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
C=CCC1=C(C(=C(C(=C1F)F)F)F)F
Refractive Index 1.427
Density 1.358
Boiling Point 148°C to 149°C
Quantity 25g
Flash Point 42°C (108°F)
UN Number UN1993
Beilstein 1971209
Formula Weight 208.13
Percent Purity 98%
Chemical Name or Material Allylpentafluorobenzene

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 217-083-6

TSCA : No

Recommended Storage : Ambient temperatures