Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylsuccinic Anhydride 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
7539-12-0
C7H8O3
140.138
MFCD00085227
WUMMIJWEUDHZCL-UHFFFAOYSA-N
4-Pentene-1,2-dicarboxylic Anhydride, (2-Propenyl)butanedioic Anhydride
111009
3-prop-2-enyloxolane-2,5-dione
C=CCC1CC(=O)OC1=O
7539-12-0
C7H8O3
140.138
MFCD00085227
WUMMIJWEUDHZCL-UHFFFAOYSA-N
4-Pentene-1,2-dicarboxylic Anhydride, (2-Propenyl)butanedioic Anhydride
111009
3-prop-2-enyloxolane-2,5-dione
C=CCC1CC(=O)OC1=O
Boiling Point 143°C
Quantity 25g
Color Yellow
UN Number 2810
Formula Weight 140.14
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material Allylsuccinic Anhydride

EINECSNumber : (5)-7013

RTECSNumber : WN1050000

TSCA : No