Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Allyltri-n-butyltin, 97%

Click to view available options
Quantity:
5g
25g
24850-33-7
C15H32Sn
331.131
MFCD00010346
YLGRTLMDMVAFNI-UHFFFAOYSA-N
allyltributyltin,allyltributylstannane,allyltri-n-butyltin,stannane, tributyl-2-propenyl,tributyl prop-2-en-1-yl stannane,allyl tributyl stannane,unii-aor8od4103,tributyl-2-propenylstannane,tri-n-butyl-2-propenylstannane,allyl tributyltin
90628
tributyl(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C
24850-33-7
C15H32Sn
331.131
MFCD00010346
YLGRTLMDMVAFNI-UHFFFAOYSA-N
allyltributyltin,allyltributylstannane,allyltri-n-butyltin,stannane, tributyl-2-propenyl,tributyl prop-2-en-1-yl stannane,allyl tributyl stannane,unii-aor8od4103,tributyl-2-propenylstannane,tri-n-butyl-2-propenylstannane,allyl tributyltin
90628
tributyl(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C
Applications
Allyltri-n-butyltin acts as an allylation reagent for aldehydes catalyzed by chiral Lewis acids and ytterbium(III) trifluoromethanesulfonate. It is also used to prepare homoallylic alcohols with trichloro-1,3,5-triazene. Further, it plays an important role for tetrakis(triphenylphosphine)palladium(0) catalyzed coupling reaction with iodoquinones and allyl acetates.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
Boiling Point 100°C to 102°C (0.5mmHg)
Quantity 5g
Flash Point 103°C (217°F)
Refractive Index 1.486
Density 1.079
Sensitivity Air sensitive
UN Number UN2788
Beilstein 3588340
Solubility Information Immiscible with water.
Formula Weight 331.12
Percent Purity 97%
Chemical Name or Material Allyltri-n-butyltin
Show More Show Less

GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

EINECSNumber : 246-494-3

TSCA : No

Recommended Storage : Keep cold; Store under Argon