Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltributyltin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
24850-33-7
C15H32Sn
331.1
MFCD00010346
YLGRTLMDMVAFNI-UHFFFAOYSA-N
allyltributyltin,allyltributylstannane,allyltri-n-butyltin,stannane, tributyl-2-propenyl,tributyl prop-2-en-1-yl stannane,allyl tributyl stannane,unii-aor8od4103,tributyl-2-propenylstannane,tri-n-butyl-2-propenylstannane,allyl tributyltin
90628
tributyl(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C
24850-33-7
C15H32Sn
331.1
MFCD00010346
YLGRTLMDMVAFNI-UHFFFAOYSA-N
allyltributyltin,allyltributylstannane,allyltri-n-butyltin,stannane, tributyl-2-propenyl,tributyl prop-2-en-1-yl stannane,allyl tributyl stannane,unii-aor8od4103,tributyl-2-propenylstannane,tri-n-butyl-2-propenylstannane,allyl tributyltin
90628
tributyl(prop-2-enyl)stannane
CCCC[Sn](CCCC)(CCCC)CC=C
Flash Point >110°C
Specific Gravity 1.068
Color Colorless
Packaging Glass bottle
Quantity 1g
Density 1.0680g/mL
Infrared Spectrum Authentic
Refractive Index 1.4850 to 1.4870
Boiling Point 88.0°C to 92.0°C (0.2mmHg)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [CH3(CH2)3]3SnCH2CH=CH2
Beilstein 04, 1922
Fieser 09,8; 11,15; 12,21; 14,14; 16,7; 17,12
Merck Index 15, 289
Formula Weight 331.1
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Allyltributyltin
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 246-494-3