Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrimethoxysilane, 97+%, Acros Organics

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
2551-83-9
C6H14O3Si
162.26
MFCD00053865
LFRDHGNFBLIJIY-UHFFFAOYSA-N
allyltrimethoxysilane,silane, trimethoxy-2-propenyl,silane, trimethoxy-2-propen-1-yl,allyl trimethoxy silane,silane, allyltrimethoxy,allytrimethoxysilane,acmc-209gkn,trimethoxy prop-2-enyl silane,silane,trimethoxy-2-propen-1-yl
75698
trimethoxy(prop-2-enyl)silane
CO[Si](CC=C)(OC)OC
2551-83-9
C6H14O3Si
162.26
MFCD00053865
LFRDHGNFBLIJIY-UHFFFAOYSA-N
allyltrimethoxysilane,silane, trimethoxy-2-propenyl,silane, trimethoxy-2-propen-1-yl,allyl trimethoxy silane,silane, allyltrimethoxy,allytrimethoxysilane,acmc-209gkn,trimethoxy prop-2-enyl silane,silane,trimethoxy-2-propen-1-yl
75698
trimethoxy(prop-2-enyl)silane
CO[Si](CC=C)(OC)OC
Color Colorless
Infrared Spectrum Authentic
Boiling Point 146°C
Refractive Index 1.4040 to 1.406
Density 0.9700g/mL
Flash Point 46°C
Specific Gravity 0.97
Packaging Glass bottle
Quantity 5mL
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97% min. (GC)
Linear Formula H2C=CHCH2Si(OCH3)3
Solubility Information Solubility in water: reacts
Formula Weight 162.26
Physical Form Liquid
Percent Purity 97+%
Chemical Name or Material Allyltrimethoxysilane
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-855-8