Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrimethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
762-72-1
C6H14Si
114.27
MFCD00008635
HYWCXWRMUZYRPH-UHFFFAOYSA-N
allyltrimethylsilane,silane, trimethyl-2-propenyl,trimethylallylsilane,silane, allyltrimethyl,allyl trimethylsilane,unii-8b84c337vf,ccris 2649,allyl trimethyl silane,3-trimethylsilyl propene,allytrimethylsilane
69808
trimethyl(prop-2-enyl)silane
C[Si](C)(C)CC=C
762-72-1
C6H14Si
114.27
MFCD00008635
HYWCXWRMUZYRPH-UHFFFAOYSA-N
allyltrimethylsilane,silane, trimethyl-2-propenyl,trimethylallylsilane,silane, allyltrimethyl,allyl trimethylsilane,unii-8b84c337vf,ccris 2649,allyl trimethyl silane,3-trimethylsilyl propene,allytrimethylsilane
69808
trimethyl(prop-2-enyl)silane
C[Si](C)(C)CC=C
Density 0.7190g/mL
Flash Point 7°C
Boiling Point 85°C
Specific Gravity 0.719
Refractive Index 1.4060 to 1.408
Quantity 50mL
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula H2C=CHCH2Si(CH3)3
Fieser 10,6; 11,16; 12,23; 13,11; 14,18; 15,8
Merck Index 15, 290
Solubility Information Solubility in water: insoluble
Formula Weight 114.27
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Allyltrimethylsilane, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-104-5

TSCA : TSCA