Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrimethylsilane, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
250mL
Packaging:
Glass bottle
762-72-1
C6H14Si
114.27
HYWCXWRMUZYRPH-UHFFFAOYSA-N
allyltrimethylsilane,silane, trimethyl-2-propenyl,trimethylallylsilane,silane, allyltrimethyl,allyl trimethylsilane,unii-8b84c337vf,ccris 2649,allyl trimethyl silane,3-trimethylsilyl propene,allytrimethylsilane
69808
trimethyl(prop-2-enyl)silane
C[Si](C)(C)CC=C
762-72-1
C6H14Si
114.27
HYWCXWRMUZYRPH-UHFFFAOYSA-N
allyltrimethylsilane,silane, trimethyl-2-propenyl,trimethylallylsilane,silane, allyltrimethyl,allyl trimethylsilane,unii-8b84c337vf,ccris 2649,allyl trimethyl silane,3-trimethylsilyl propene,allytrimethylsilane
69808
trimethyl(prop-2-enyl)silane
C[Si](C)(C)CC=C
Refractive Index 1.4060 to 1.4080
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 0.719
Color Colorless
Density 0.7190g/mL
Quantity 10mL
Boiling Point 85.0°C
Flash Point 7°C
Assay Percent Range 99%
Linear Formula H2C=CHCH2Si(CH3)3
Fieser 10,6; 11,16; 12,23; 13,11; 14,18; 15,8
Merck Index 15,29
Solubility Information Solubility in water: insoluble
Formula Weight 114.27
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Allyltrimethylsilane
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 212-104-5