Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltriphenylphosphonium Chloride 98.0+%, TCI America™

Click to view available options
Quantity:
25g
18480-23-4
C21H20ClP
338.815
MFCD00031542
FKMJROWWQOJRJX-UHFFFAOYSA-M
allyltriphenylphosphonium chloride,allyl triphenylphosphonium chloride,triphenyl prop-2-enyl phosphanium chloride,triphenyl prop-2-en-1-yl phosphanium chloride,acmc-1buz2,allyltriphenylphosphoniumchloride,triphenyl prop-2-enyl phosphonium chloride,phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
2734120
triphenyl(prop-2-enyl)phosphanium;chloride
C=CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
18480-23-4
C21H20ClP
338.815
MFCD00031542
FKMJROWWQOJRJX-UHFFFAOYSA-M
allyltriphenylphosphonium chloride,allyl triphenylphosphonium chloride,triphenyl prop-2-enyl phosphanium chloride,triphenyl prop-2-en-1-yl phosphanium chloride,acmc-1buz2,allyltriphenylphosphoniumchloride,triphenyl prop-2-enyl phosphonium chloride,phosphonium,triphenyl-2-propen-1-yl-, chloride 1:1
2734120
triphenyl(prop-2-enyl)phosphanium;chloride
C=CC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
Quantity 25g
Melting Point 230°C
Color White-Yellow
Formula Weight 338.81
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material Allyltriphenylphosphonium Chloride