Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Almond Oil, Sweet, NF, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
8007-69-0
8007-69-0

AL215, 8007-69-0

Spectrum™ Almond Oil, Sweet, NF is a natural, versatile and effective emollient known for its stability and high level of the "skin-nourishing" Linoleic Acid. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 100%