Learn More
Selleck Chemical LLC Almonertinib2mg
SDP
Supplier:  Selleck Chemical LLC S88172MG
Encompass_Preferred
Almonertinib (Aumolertinib, HS-10296, Ameile) is a small molecule inhibitor of EGFR-activating mutations and T790M-resistant mutation with limited activity against wild-type EGFR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.