Learn More
Ansell MICROFLEX™ Aloe Sense™ N96 Nitrile Gloves
Powder free, nitrile exam glove provides reliable protection and extra sensitivity with soothing aloe and chamomile inner coating.
$18.31
Specifications
Applications Laboratory, Healthcare
AQL 1.5
Certifications/Compliance ASTM D6319, FDA21 CFR 177-2600, ISO 13485:2003
Thickness (English) 3.5 mil
Thickness (Metric) 0.09 mm
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-170-824
View Documents
Ansell
N960
X-Small
Pack of 100 for $18.31
Case of 10 PK for $183.06
Add to cart
19-170-825
View Documents
Ansell
N961
Small
Pack of 100 for $18.31
Case of 10 PK for $183.06
Edge
Add to cart
19-170-826
View Documents
Ansell
N962
Medium
Pack of 100 for $18.31
Case of 10 PK for $183.06
Add to cart
19-170-827
View Documents
Ansell
N963
Large
Pack of 100 for $18.31
Case of 10 PK for $183.06
Add to cart
19-170-828
View Documents
Ansell
N964
X-Large
Pack of 100 for $18.31
Case of 10 PK for $183.06
Add to cart
Description
Description
  • Extra soft nitrile exam glove
  • Tactile sensitivity and comfort
  • Soothing aloe helps protect the hands from drying and cracking
  • Fully textured for a secure grip
  • Recommended for standard patient examinations, laboratory analysis, sample taking/processing and light assembly tasks
Specifications
Specifications
Laboratory, Healthcare
ASTM D6319, FDA21 CFR 177-2600, ISO 13485:2003
0.09 mm
14 MPa
0.12 mm
0.09 mm
1.5
3.5 mil
Partially Textured
4.7 mil
3.5 mil