Learn More
Abnova™ ALOX15 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00000246T01
Catalog No. 89-933-214
Add to cart
Description
Description
Specifications
Specifications
293T
BC029032.1
pCMV-ALOX15 full-length
100 μL
Human
ALOX15 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 74.8kDa
Transient overexpression cell lysate was tested with anti-ALOX15 antibody (Mfr. No. H00000246-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot