Learn More
ALOX15 Mouse anti-Human, Clone: OTI7H6, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504358
Catalog No. 50-167-5072
Add to cart
Description
Description
ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
Specifications
Specifications
ALOX15
Monoclonal
Unconjugated
ALOX15
Liquid
ALOX15
Full length human recombit protein of human ALOX15 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI7H6
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P16050
12-LOX, 15-LOX, 15-LOX-1, LOG15
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
246