Learn More
ALOX5 Mouse anti-Human, Clone: OTI1A1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806885
Catalog No. 50-166-8029
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT ). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma, fibrosis and atherosclerosis. Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO. Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation.
Specifications
Specifications
ALOX5
Monoclonal
Unconjugated
ALOX5
Lyophilized
ALOX5
Human recombit protein fragment corresponding to amino acids 18-364 of human ALOX5 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1A1
PBS with 8% trehalose and no preservative; pH 7.3
P09917
5-LO, 5-LOX, 5LPG, LOG5
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
240