Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ alpha 1 Mannosidase 1A Knockout HeLa Cell Lysate

This product is for research use only and is not approved for use in humans or in clinical diagnosis.

Manufacturer:  Novus Biologicals™ NBP266180

Catalog No. NBP266180


Add to cart

Description

Description

The alpha 1 Mannosidase 1A Knockout HeLa Cell Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications

Specifications

LYOPH
Human
0.1 mg cell homogenate lyophilized in RIPA buffer made with double-knockout cell lines
alpha 1 Mannosidase 1A
Western Blot
2 Vials
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit